(1R,2R)-1-Amino-2-indanol

Lieferant: Apollo Scientific

Synonyme: (1R,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol, (1R,2R)-(-)-1-Amino-2-hydroxyindan, (1R,2R)-1-Aminoindan-2-ol

OR55290-1G OR55290-5G
APOSOR55290-1GEA 59.9 EUR
APOSOR55290-1G APOSOR55290-5G
(1R,2R)-1-Amino-2-indanol
(1R,2R)-(-)-1-Amino-2-indanol
Formel: C₉H₁₁NO
Molecular Weight: 149.19 g/mol
Schmelzpunkt: 142…146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08275447
CAS-Nummer: 163061-73-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR