Ethyl-2,4,6-trimethylphenylacetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl mesitylacetate

OR55298-10G OR55298-2G
APOSOR55298-10GEA 292 EUR
APOSOR55298-10G APOSOR55298-2G
Ethyl-2,4,6-trimethylphenylacetate
Ethyl-2,4,6-trimethylphenylacetate
Formel: C₁₃H₁₈O₂
Molecular Weight: 206,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051916
CAS-Nummer: 5460-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR