3-Chlor-(4-ethoxycarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: (3-Chloro-4-(ethoxycarbonyl)phenyl)boronic acid, 3-Chloro-4-(ethoxycarbonyl)phenylboronic acid

OR5532-1G OR5532-5G
APOSOR5532-1GEA 97.3 EUR
APOSOR5532-1G APOSOR5532-5G
3-Chlor-(4-ethoxycarbonyl)phenylboronsäure 98%
3-Chlor-(4-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₄
Molecular Weight: 228,4373 g/mol
Schmelzpunkt: 122…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659875
CAS-Nummer: 850568-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR