5-Chlor-2-methoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-Chloro-2-methoxyphenyl)boronic acid, 5-Chloro-2-methoxybenzeneboronic acid
OR5534-25G
OR5534-5G
APOSOR5534-25GEA
42.2
EUR
APOSOR5534-25G
APOSOR5534-5G
5-Chlor-2-methoxyphenylboronsäure 98%
5-Chlor-2-methoxyphenylboronsäure
Formel:
C₇H₈BClO₃ Molecular Weight: 186,40 g/mol Schmelzpunkt: 134…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318966 CAS-Nummer: 89694-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...