Allylchlordimethylsilan 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Allyldimethylchlorsilan, ADMCS
OR55393-25G
OR55393-5G
APOSOR55393-25GEA
256
EUR
APOSOR55393-25G
APOSOR55393-5G
Allylchlordimethylsilan 95%
Allylchlordimethylsilan
Formel:
C₅H₁₁ClSi Molecular Weight: 134.68 g/mol Siedepunkt: 111…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000506 CAS-Nummer: 4028-23-3 EINECS: 223-706-2 UN: 2985 ADR: 3+8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...