6-Nitroanthranilic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-6-nitrobenzoic Acid

OR55396-1G OR55396-25G OR55396-5G
APOSOR55396-1GEA 21.9 EUR
APOSOR55396-1G APOSOR55396-25G APOSOR55396-5G
6-Nitroanthranilic acid
6-Nitroanthranilic acid
Formel: C₇H₆N₂O₄
Molecular Weight: 182,14 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00034785
CAS-Nummer: 50573-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR