5-Chloro-1H-pyrazolo[3,4-c]pyridine 97%

Lieferant: Apollo Scientific
Achtung

OR55452-5G
APOSOR55452-5GEA 139 EUR
APOSOR55452-5G
5-Chloro-1H-pyrazolo[3,4-c]pyridine 97%
5-Chloro-1H-pyrazolo[3,4-c]pyridine
Formel: C₆H₄ClN₃
Molecular Weight: 153,5691 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05663983
CAS-Nummer: 76006-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR