4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Chloro-4-(methoxycarbonyl)benzeneboronic acid, pinacol ester

OR5549-1G OR5549-5G
APOSOR5549-1GEA 127 EUR
APOSOR5549-1G APOSOR5549-5G
4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester 95%
4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester
Formel: C₁₄H₁₈BClO₄
Molecular Weight: 296,56 g/mol
Schmelzpunkt: 48…51 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659928
CAS-Nummer: 334018-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR