2-Bromo-N,N-dimethylacetamide 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR55573-25G
OR55573-5G
OR55573-100G
APOSOR55573-25GEA
143
EUR
APOSOR55573-25G
APOSOR55573-5G
APOSOR55573-100G
2-Bromo-N,N-dimethylacetamide 98%
2-Bromo-N,N-dimethylacetamide
Formel:
C₄H₈BrNO Molecular Weight: 166.02 g/mol Siedepunkt: 63…65 °C/1mmHg Dichte: 1.5412 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD12026683 CAS-Nummer: 5468-77-9 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...