1-tert-Butylimidazole 98+%
Lieferant: Apollo Scientific
|
Achtung
|
OR55585-25G
OR55585-5G
APOSOR55585-25GEA
495
EUR
APOSOR55585-25G
APOSOR55585-5G
1-tert-Butylimidazole 98+%
1-tert-Butylimidazole
Formel:
C₇H₁₂N₂ Molecular Weight: 124,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19441219 CAS-Nummer: 45676-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...