(4-Methoxybenzyl)hydrazine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR55590-100G
APOSOR55590-100GEA 88.6 EUR
APOSOR55590-100G
(4-Methoxybenzyl)hydrazine hydrochloride 97%
(4-Methoxybenzyl)hydrazine hydrochloride
Formel: C₈H₁₃ClN₂O
Molecular Weight: 188,6546 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01722406
CAS-Nummer: 2011-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR