1,3-Dibrom-2-iodbenzol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dibromoiodobenzene

OR55629-1G OR55629-5G
APOSOR55629-1GEA 26.1 EUR
APOSOR55629-1G APOSOR55629-5G
1,3-Dibrom-2-iodbenzol
1,3-Dibromo-2-iodobenzene
Formel: C₆H₃Br₂I
Molecular Weight: 361,80 g/mol
Schmelzpunkt: 100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192666
CAS-Nummer: 19821-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR