5-Hydroxynicotinsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Hydroxypyridin-3-carbonsäure, 5-Hydroxy-3-pyridincarbonsäure

OR5565-25G OR5565-5G
APOSOR5565-25GEA 24.8 EUR
APOSOR5565-25G APOSOR5565-5G
5-Hydroxynicotinsäure 97%
5-Hydroxynicotinsäure
Formel: C₆H₅NO₃
Molecular Weight: 139,11 g/mol
Schmelzpunkt: 300…304 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00129117
CAS-Nummer: 27828-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR