7-Chloro-1-tetralone 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Chloro-3,4-dihydronaphthalen-1(2H)-one, 7-Chloro-3,4-dihydro-1(2H)-naphthalenone

OR55702-1G OR55702-25G OR55702-5G
APOSOR55702-1GEA 21.9 EUR
APOSOR55702-1G APOSOR55702-25G APOSOR55702-5G
7-Chloro-1-tetralone 99%
7-Chloro-1-tetralone
Formel: C₁₀H₉ClO
Molecular Weight: 180,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01928386
CAS-Nummer: 26673-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR