8-hydroxyquinoline-2-carbonitrile 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Hydroxyquinoline-2-carbonitrile

OR55719-1G OR55719-250MG OR55719-5G
APOSOR55719-1GEA 109 EUR
APOSOR55719-1G APOSOR55719-250MG APOSOR55719-5G
8-hydroxyquinoline-2-carbonitrile 99%
8-hydroxyquinoline-2-carbonitrile
Formel: C₁₀H₆N₂O
Molecular Weight: 170.171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00216717
CAS-Nummer: 6759-78-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR