2-Hydroxy-3-nitro-5-picolin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methyl-3-nitro-2-pyridon, 2-Hydroxy-5-methyl-3-nitropyridin
OR5572-5G
OR5572-1G
APOSOR5572-5GEA
27.7
EUR
APOSOR5572-5G
APOSOR5572-1G
2-Hydroxy-3-nitro-5-picolin 98%
2-Hydroxy-3-nitro-5-picolin
Formel:
C₆H₆N₂O₃ Molecular Weight: 154.13 g/mol Schmelzpunkt: 179 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02070021 CAS-Nummer: 7464-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...