3-Brom-2-chlor-5-nitropyridin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR5575-100G
APOSOR5575-100GEA
161
EUR
APOSOR5575-100G
3-Brom-2-chlor-5-nitropyridin 98%
3-Bromo-2-chloro-5-nitropyridine
Formel:
C₅H₂BrClN₂O₂ Schmelzpunkt: 67…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233989 CAS-Nummer: 5470-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...