2-Formyl-5-methoxyphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methoxy-2-formylphenylboronic acid

OR5581-1G OR5581-5G OR5581-100MG
APOSOR5581-1GEA 125 EUR
APOSOR5581-1G APOSOR5581-5G APOSOR5581-100MG
2-Formyl-5-methoxyphenylboronsäure
2-Formyl-5-methoxyphenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.9657 g/mol
Schmelzpunkt: 155…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03001335
CAS-Nummer: 40138-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR