Ethyl-2,4-dihydroxy-6-methylnicotinat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylate, 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylic acid ethyl ester, Ethyl 2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, Ethyl 2,4-Dihydroxy-6-methylnicotinate, 2,4-Dihydroxy-6-methylnicotinic acid ethyl ester

OR5584-1G OR5584-25G
APOSOR5584-1GEA 21.9 EUR
APOSOR5584-1G APOSOR5584-25G
Ethyl-2,4-dihydroxy-6-methylnicotinat 97%
Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate
Formel: C₉H₁₁NO₄
Molecular Weight: 197,1900 g/mol
Schmelzpunkt: 210…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02929397
CAS-Nummer: 70254-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR