1H-Imidazol-4-carbonsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Imidazole-4-carboxylic acid
OR5585-1G
OR5585-25G
OR5585-5G
APOSOR5585-1GEA
21.9
EUR
APOSOR5585-1G
APOSOR5585-25G
APOSOR5585-5G
1H-Imidazol-4-carbonsäure 95%
1H-Imidazole-4-carboxylic acid
Formel:
C₄H₄N₂O₂ Schmelzpunkt: 264…266 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082203 CAS-Nummer: 1072-84-0 EINECS: 214-017-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...