2-(4-Bromphenylsulfonamido)essigsäure 98%

Lieferant: Apollo Scientific

OR5595-25G OR5595-5G
APOSOR5595-25GEA 274 EUR
APOSOR5595-25G APOSOR5595-5G
2-(4-Bromphenylsulfonamido)essigsäure 98%
2-(4-Bromphenylsulfonamido)essigsäure
Formel: C₈H₈BrNO₄S
Schmelzpunkt: 194…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01935943
CAS-Nummer: 13029-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR