2-Bromo-5-hydroxymethylthiazole 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromothiazole-5-methanol
OR5599-1G
APOSOR5599-1GEA
109
EUR
APOSOR5599-1G
2-Bromo-5-hydroxymethylthiazole 98%
2-Bromo-5-hydroxymethylthiazole
Formel:
C₄H₄BrNOS Molecular Weight: 194,06 g/mol Schmelzpunkt: 46…48 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04115733 CAS-Nummer: 687636-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...