2-Amino-4-(4-chlorphenyl)thiazol 98%

Lieferant: Apollo Scientific

Synonyme: 4-(4-Chlorophenyl)thiazol-2-amine

OR5604-10G OR5604-25G
APOSOR5604-10GEA 94.5 EUR
APOSOR5604-10G APOSOR5604-25G
2-Amino-4-(4-chlorphenyl)thiazol 98%
2-Amino-4-(4-chlorophenyl)thiazole
Formel: C₉H₇ClN₂S
Molecular Weight: 210,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047058
CAS-Nummer: 2103-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR