1-Aminobenzotriazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR5609-1G
OR5609-250MG
OR5609-5G
APOSOR5609-1GEA
109
EUR
APOSOR5609-1G
APOSOR5609-250MG
APOSOR5609-5G
1-Aminobenzotriazol 97%
1-Aminobenzotriazole
Formel:
C₆H₆N₄ Schmelzpunkt: 81…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00132902 CAS-Nummer: 1614-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...