Ethyl-2-amino-5-oxazolcarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-aminooxazole-5-carboxylate
OR5611-1G
OR5611-250MG
APOSOR5611-1GEA
26.1
EUR
APOSOR5611-1G
APOSOR5611-250MG
Ethyl-2-amino-5-oxazolcarboxylat
Ethyl-2-amino-5-oxazolcarboxylat
Formel:
C₆H₈N₂O₃ Molecular Weight: 156,1393 g/mol Schmelzpunkt: 151…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657433 CAS-Nummer: 113853-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...