Methyl-2-amino-5-bromothiazole-4-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-amino-5-bromothiazole-4-carboxylate
OR5615-1G
OR5615-5G
APOSOR5615-1GEA
21.9
EUR
APOSOR5615-1G
APOSOR5615-5G
Methyl-2-amino-5-bromothiazole-4-carboxylate 95%
Methyl-2-amino-5-bromothiazole-4-carboxylate
Formel:
C₅H₅BrN₂O₂S Molecular Weight: 237,08 g/mol Schmelzpunkt: 157…159 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00137990 CAS-Nummer: 850429-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...