3-Amino-5-tert-butylisoxazol 97%

Lieferant: Apollo Scientific
Achtung

OR5616-100G OR5616-25G OR5616-5G
APOSOR5616-100GEA 321 EUR
APOSOR5616-100G APOSOR5616-25G APOSOR5616-5G
3-Amino-5-tert-butylisoxazol 97%
3-Amino-5-tert-butylisoxazol
Formel: C₇H₁₂N₂O
Molecular Weight: 140,19 g/mol
Schmelzpunkt: 110…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00055620
CAS-Nummer: 55809-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR