1-(2-Methoxyphenyl)-2-thioharnstoff
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Methoxyphenylthioharnstoff, N-(2-Methoxyphenyl)thioharnstoff
OR5634-1G
OR5634-5G
APOSOR5634-1GEA
21.9
EUR
APOSOR5634-1G
APOSOR5634-5G
1-(2-Methoxyphenyl)-2-thioharnstoff
1-(2-Methoxyphenyl)-2-thioharnstoff
Formel:
(C₆H₄(OCH₃))NC(S)NH₂ Molecular Weight: 182,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022162 CAS-Nummer: 1516-37-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...