Benzylhydrazine monohydrochloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Benzylhydrazine hydrochloride
OR5636-25G
APOSOR5636-25GEA
75.4
EUR
APOSOR5636-25G
Benzylhydrazine monohydrochloride
Benzylhydrazine monohydrochloride
Formel:
C₇H₁₀N₂ Molecular Weight: 158,63 g/mol Schmelzpunkt: 110…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01722685 CAS-Nummer: 1073-62-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...