4-tert-Butylthiazol-2-ylamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-(tert-butyl)-1,3-thiazole
OR5658-1G
OR5658-5G
APOSOR5658-1GEA
32.1
EUR
APOSOR5658-1G
APOSOR5658-5G
4-tert-Butylthiazol-2-ylamine
4-tert-Butylthiazol-2-ylamine
Formel:
C₇H₁₂N₂S Molecular Weight: 156,25 g/mol Schmelzpunkt: 99…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051644 CAS-Nummer: 74370-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...