2-Indolcarbaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Indol-2-carboxaldehyd
OR5662-5G
OR5662-1G
OR5662-250MG
APOSOR5662-5GEA
102
EUR
APOSOR5662-5G
APOSOR5662-1G
APOSOR5662-250MG
2-Indolcarbaldehyd
2-Indolcarbaldehyd
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Schmelzpunkt: 138…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001425 CAS-Nummer: 19005-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...