(Pyridin-2-ylmethyl)(pyridin-3-ylmethyl)amine

Lieferant: Apollo Scientific
Achtung

OR5671-1G OR5671-250MG
APOSOR5671-1GEA 184 EUR
APOSOR5671-1G APOSOR5671-250MG
(Pyridin-2-ylmethyl)(pyridin-3-ylmethyl)amine
(Pyridin-2-ylmethyl)(pyridin-3-ylmethyl)amine
Formel: C₁₂H₁₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02656617
CAS-Nummer: 128802-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR