3-Benzyl-7-hydroxy-4-methyl-2H-chromen-2-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Benzyl-7-hydroxy-4-methylcoumarin

OR5674-1G OR5674-500MG OR5674-250MG
APOSOR5674-1GEA 35 EUR
APOSOR5674-1G APOSOR5674-500MG APOSOR5674-250MG
3-Benzyl-7-hydroxy-4-methyl-2H-chromen-2-one
3-Benzyl-7-hydroxy-4-methyl-2H-chromen-2-one
Formel: C₁₇H₁₄O₃
Molecular Weight: 266.3 g/mol
Schmelzpunkt: 232…234 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00047643
CAS-Nummer: 86-44-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR