1-(4-Bromphenylsulfonyl)pyrrolidin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-N-pyrrolidin-1-ylbenzenesulphonamide, 1-((4-Bromophenyl)sulphonyl)pyrrolidine
OR5699-1G
OR5699-5G
APOSOR5699-1GEA
125
EUR
APOSOR5699-1G
APOSOR5699-5G
1-(4-Bromphenylsulfonyl)pyrrolidin 98%
1-(4-Bromphenylsulfonyl)pyrrolidin
Formel:
C₁₀H₁₂BrNO₂S Molecular Weight: 290,1768 g/mol Schmelzpunkt: 80…84 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00584577 CAS-Nummer: 136350-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...