1-((4-Bromphenyl)sulfonyl)piperidin 98%
Lieferant: Apollo Scientific
OR5704-1G
OR5704-5G
APOSOR5704-1GEA
46.4
EUR
APOSOR5704-1G
APOSOR5704-5G
1-((4-Bromphenyl)sulfonyl)piperidin 98%
1-((4-Bromphenyl)sulfonyl)piperidin
Formel:
C₁₁H₁₄BrNO₂S Molecular Weight: 304.2034 g/mol Schmelzpunkt: 91…93 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00194753 CAS-Nummer: 834-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...