4-(4-Bromphenylsulfonyl)morpholin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-((4-Bromophenyl)sulphonyl)morpholine

OR5705-1G OR5705-25G OR5705-5G
APOSOR5705-1GEA 101 EUR
APOSOR5705-1G APOSOR5705-25G APOSOR5705-5G
4-(4-Bromphenylsulfonyl)morpholin 98%
4-(4-Bromphenylsulfonyl)morpholin
Formel: C₁₀H₁₂BrNO₃S
Molecular Weight: 306,1800 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00194739
CAS-Nummer: 834-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR