4-Iod-2,5-xylenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,5-Dimethyl-4-iodphenol, 2-Hydroxy-5-iod-4-xylol, 5-Hydroxy-2-iod-4-xylol
OR5710-1G
OR5710-5G
APOSOR5710-1GEA
154
EUR
APOSOR5710-1G
APOSOR5710-5G
4-Iod-2,5-xylenol 98%
4-Iod-2,5-xylenol
Formel:
C₈H₉IO Molecular Weight: 248,06 g/mol Schmelzpunkt: 95…97 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00219922 CAS-Nummer: 114971-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...