5-(Boc-amino)-2-brompyridin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl (6-bromopyridin-3-yl)carbamate, 5-Amino-2-bromopyridine, 5-BOC protected
OR5734-1G
OR5734-250MG
APOSOR5734-1GEA
29
EUR
APOSOR5734-1G
APOSOR5734-250MG
5-(Boc-amino)-2-brompyridin 95%
5-(Boc-amino)-2-brompyridin
Formel:
C₁₀H₁₃BrN₂O₂ Molecular Weight: 273.13 g/mol Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07363808 CAS-Nummer: 218594-15-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...