3-Brom-5-nitrobenzoesäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR5735-1G
OR5735-25G
OR5735-5G
APOSOR5735-1GEA
21.9
EUR
APOSOR5735-1G
APOSOR5735-25G
APOSOR5735-5G
3-Brom-5-nitrobenzoesäure 95%
3-Bromo-5-nitrobenzoic acid
Formel:
C₇H₄BrNO₄ Molecular Weight: 246,02 g/mol Schmelzpunkt: 159…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00100098 CAS-Nummer: 6307-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...