1-Methylpiperidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR5741-5G OR5741-1G OR5741-250MG
APOSOR5741-5GEA 230 EUR
APOSOR5741-5G APOSOR5741-1G APOSOR5741-250MG
1-Methylpiperidine-3-carboxylic acid
1-Methylpiperidine-3-carboxylic acid
Formel: C₇H₁₃NO₂
Schmelzpunkt: 162…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08059558
CAS-Nummer: 5657-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR