1-(Piperidin-3-ylcarbonyl)piperidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3'-Carbonyldi(piperidine)

OR5744-1G OR5744-250MG
APOSOR5744-1GEA 132 EUR
APOSOR5744-1G APOSOR5744-250MG
1-(Piperidin-3-ylcarbonyl)piperidine
1-(Piperidin-3-ylcarbonyl)piperidine
Formel: C₁₁H₂₀N₂O
Molecular Weight: 196,29 g/mol
Siedepunkt: 120…125 °C/0,3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01716373
CAS-Nummer: 40576-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR