2-Ethoxy-4-nitrobenzoesäure

Lieferant: Apollo Scientific
Achtung

OR5746-25G OR5746-5G OR5746-100G
APOSOR5746-25GEA 250 EUR
APOSOR5746-25G APOSOR5746-5G APOSOR5746-100G
2-Ethoxy-4-nitrobenzoesäure
2-Ethoxy-4-nitrobenzoic acid
Formel: C₉H₉NO₅
Molecular Weight: 211,17 g/mol
Schmelzpunkt: 148…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683865
CAS-Nummer: 2486-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR