4-Carbamoyl-3-chlorobenzeneboronic acid 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Carbamoyl-3-chlorophenyl)boronic acid, 4-(Aminocarbonyl)-3-chlorobenzeneboronic acid, 4-Aminocarbonyl-3-chlorophenylboronic acid
OR5761-1G
OR5761-5G
APOSOR5761-1GEA
193
EUR
APOSOR5761-1G
APOSOR5761-5G
4-Carbamoyl-3-chlorobenzeneboronic acid 96%
4-Carbamoyl-3-chlorobenzeneboronic acid
Formel:
C₇H₇BClNO₃ Molecular Weight: 199,40 g/mol Schmelzpunkt: 152…158 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363777 CAS-Nummer: 850589-52-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...