4-(4-Carboxypiperidin-1-carbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Boronobenzoyl)piperidine-4-carboxylic acid, 4-(1-Carbonyl-4-carboxypiperidine)benzeneboronic acid

OR5765-1G OR5765-5G
APOSOR5765-1GEA 181 EUR
APOSOR5765-1G APOSOR5765-5G
4-(4-Carboxypiperidin-1-carbonyl)phenylboronsäure 98%
4-(4-Carboxypiperidin-1-carbonyl)phenylboronsäure
Formel: C₁₃H₁₆BNO₅
Molecular Weight: 277.0800 g/mol
Schmelzpunkt: 110…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363784
CAS-Nummer: 850593-02-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR