3-Acetamido-5-brombenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR5768-1G
OR5768-25G
OR5768-5G
APOSOR5768-1GEA
108
EUR
APOSOR5768-1G
APOSOR5768-25G
APOSOR5768-5G
3-Acetamido-5-brombenzoesäure 98%
3-Acetamido-5-brombenzoesäure
Formel:
C₉H₈BrNO₃ Schmelzpunkt: 282…284 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01098204 CAS-Nummer: 78238-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...