2-Methoxynicotinonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Cyano-2-methoxypyridine, 2-Methoxynicotinonitrile

OR5771-1G OR5771-5G OR5771-10G
APOSOR5771-1GEA 23.2 EUR
APOSOR5771-1G APOSOR5771-5G APOSOR5771-10G
2-Methoxynicotinonitril 98%
2-Methoxynicotinonitril
Formel: C₇H₆N₂O
Molecular Weight: 134,14 g/mol
Schmelzpunkt: 58…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363804
CAS-Nummer: 7254-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR