4-Chlor-2-ethoxy-1-nitrobenzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR5777-1G
OR5777-5G
APOSOR5777-1GEA
80
EUR
APOSOR5777-1G
APOSOR5777-5G
4-Chlor-2-ethoxy-1-nitrobenzol 98%
4-Chlor-2-ethoxy-1-nitrobenzol
Formel:
C₈H₈ClNO₃ Schmelzpunkt: 52…54 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02625956 CAS-Nummer: 29604-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...