2-Chlor-4-(diethylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

OR5779-1G OR5779-5G
APOSOR5779-1GEA 245 EUR
APOSOR5779-1G APOSOR5779-5G
2-Chlor-4-(diethylcarbamoyl)phenylboronsäure 98%
2-Chlor-4-(diethylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₅BClNO₃
Molecular Weight: 255.5 g/mol
Schmelzpunkt: 148…152 °C
Lagertemperatur: Kühlschrank
MDL: MFCD34824970
CAS-Nummer: 850589-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR