(2,4-Dichlorphenyl)essigsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
DCAA
OR5793-100G
OR5793-25G
OR5793-500G
APOSOR5793-100GEA
36.4
EUR
APOSOR5793-100G
APOSOR5793-25G
APOSOR5793-500G
(2,4-Dichlorphenyl)essigsäure
(2,4-Dichlorphenyl)essigsäure
Formel:
(C₆H₃Cl₂)CH₂COOH Molecular Weight: 205.04 g/mol Siedepunkt: 316 Schmelzpunkt: 128…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004318 CAS-Nummer: 19719-28-9 EINECS: 243-248-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...