3-Methoxy-4-(methoxycarbonyl)phenylboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-carboxy-3-methoxyphenyl boronic acid methyl ester, 3-Methoxy-4-methoxycarbonylphenylboronic acid, Benzoic acid, 4-borono-2-methoxy-, methyl ester, 3-Methoxy-4-(methoxycarbonyl)benzeneboronic acid

OR5799-1G OR5799-5G
APOSOR5799-1GEA 21.9 EUR
APOSOR5799-1G APOSOR5799-5G
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid
Formel: C₉H₁₁BO₅
Molecular Weight: 209,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366483
CAS-Nummer: 603122-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR